COM448679430 Combi-Blocks, Inc.

4-((((9H-Fluoren-9-yl)methyl9h-fluoren-9-yl)methoxy)carbonyl)piperazine-2-carboxylic acid, HCl

  • MDL number MFCD09998239
  • Cas Number : 1352305-15-7
  • Molecular Weight : 388.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1484
Availability details for
Quantity
  • Availability
  • Add to Cart