COM448679435 Combi-Blocks, Inc.

2,3,6-Trimethoxypyridin-4-ol

  • MDL number MFCD22887394
  • Cas Number : 1414863-98-1
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1495
Availability details for
Quantity
  • Availability
  • Add to Cart