COM448679470 Combi-Blocks, Inc.

4-(2-(Methylthio)pyrimidin-4-yl)-1h-pyrazol-5-amine

  • MDL number MFCD17926250
  • Cas Number : 956722-10-4
  • Molecular Weight : 207.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1550

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1550
Availability details for
Quantity
  • Availability
  • Add to Cart