COM448679518 Combi-Blocks, Inc.

4-(2-Methoxyethylamino)-2-(methylthio)pyrimidine-5-carboxylic acid

  • MDL number MFCD11040498
  • Cas Number : 76360-92-4
  • Molecular Weight : 243.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1632

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1632
Availability details for
Quantity
  • Availability
  • Add to Cart