COM448679529 Combi-Blocks, Inc.

4-(2-Methoxyphenylsulfanyl)piperidine, HCl

  • MDL number MFCD03840149
  • Cas Number : 1170287-04-3
  • Molecular Weight : 259.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1651

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1651
Availability details for
Quantity
  • Availability
  • Add to Cart