COM448679552 Combi-Blocks, Inc.

4-(3,5-Dichloro-phenyl)-piperidine

  • MDL number MFCD03839910
  • Cas Number : 475653-05-5
  • Molecular Weight : 230.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1684
Availability details for
Quantity
  • Availability
  • Add to Cart