COM448679578 Combi-Blocks, Inc.

4-(3-Fluoro-phenyl)-piperidine

  • MDL number MFCD05189920
  • Cas Number : 104774-88-1
  • Molecular Weight : 179.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1728
Availability details for
Quantity
  • Availability
  • Add to Cart