COM448679593 Combi-Blocks, Inc.

Methyl 3,4-dihydro-2h-pyrano[2,3-b]pyridine-6-carboxylate

  • MDL number MFCD22887430
  • Cas Number : 1414864-15-5
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1753
Availability details for
Quantity
  • Availability
  • Add to Cart