COM448679613 Combi-Blocks, Inc.

4-(4,5-Dihydroimidazol-2-yl)-pyridine

  • MDL number MFCD00129053
  • Cas Number : 21381-61-3
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1783
Availability details for
Quantity
  • Availability
  • Add to Cart