COM448679631 Combi-Blocks, Inc.

1-Benzoyl-6-bromo-7-azaindole

  • MDL number MFCD09880008
  • Cas Number : 143468-12-6
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1816

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1816
Availability details for
Quantity
  • Availability
  • Add to Cart