COM448679633 Combi-Blocks, Inc.

4-(4-Bromophenoxy)-1,2-dichlorobenzene

  • MDL number MFCD09788599
  • Cas Number : 854259-28-2
  • Molecular Weight : 318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1818
Availability details for
Quantity
  • Availability
  • Add to Cart