COM448679643 Combi-Blocks, Inc.

4-(4-Bromo-phenylamino)-benzoic acid ethyl ester

  • MDL number MFCD04115310
  • Cas Number : 458550-44-2
  • Molecular Weight : 320.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1857
Availability details for
Quantity
  • Availability
  • Add to Cart