COM448679649 Combi-Blocks, Inc.

N-[2-(Aminomethyl)benzyl]-N,N-dimethylamine

  • MDL number MFCD07324744
  • Cas Number : 53369-77-0
  • Molecular Weight : 164.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1868

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1868
Availability details for
Quantity
  • Availability
  • Add to Cart