COM448679679 Combi-Blocks, Inc.

4-(4'-Cyanophenyl)-1-n-boc-aniline

  • MDL number MFCD03426381
  • Cas Number : 672309-99-8
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-1965
Availability details for
Quantity
  • Availability
  • Add to Cart