COM448679706 Combi-Blocks, Inc.

4-(4-Fluorophenyl)-4-phenylpiperidine, HCl

  • MDL number MFCD03840118
  • Cas Number : 1189673-94-6
  • Molecular Weight : 291.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2053
Availability details for
Quantity
  • Availability
  • Add to Cart