COM448679778 Combi-Blocks, Inc.

N-Cyclopropylthiourea

  • MDL number MFCD06655443
  • Cas Number : 56541-14-1
  • Molecular Weight : 116.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2277
Availability details for
Quantity
  • Availability
  • Add to Cart