COM448679794 Combi-Blocks, Inc.

5,5'-Methylenedisalicylic acid

  • MDL number MFCD00016506
  • Cas Number : 27496-82-8
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2313
Availability details for
Quantity
  • Availability
  • Add to Cart