COM448679814 Combi-Blocks, Inc.

4-(4-Methylphenyl)-4-phenylpiperidine, HCl

  • MDL number MFCD03840119
  • Cas Number : 851585-66-5
  • Molecular Weight : 287.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2341
Availability details for
Quantity
  • Availability
  • Add to Cart