COM448679831 Combi-Blocks, Inc.

4-(5-Chloromethyl-[1,2,4]oxadiazol-3-yl)-aniline

  • MDL number MFCD07369925
  • Cas Number : 6674-17-5
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2375

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2375
Availability details for
Quantity
  • Availability
  • Add to Cart