COM448679838 Combi-Blocks, Inc.

4-(6-Fluoropyridin-2-yl)piperidine-4-carbonitrile, HCl

  • MDL number MFCD11100900
  • Cas Number : 1185145-94-1
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2394
Availability details for
Quantity
  • Availability
  • Add to Cart