COM448679849 Combi-Blocks, Inc.

4-(Bromomethyl)-8-fluoroquinolin-2(1h)-one

  • MDL number MFCD18711491
  • Cas Number : 176912-29-1
  • Molecular Weight : 256.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2420
Availability details for
Quantity
  • Availability
  • Add to Cart