COM448679856 Combi-Blocks, Inc.

4-(Cyano(phenyl)methylamino)benzoic acid

  • MDL number MFCD09788590
  • Cas Number : 88170-04-1
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2437
Availability details for
Quantity
  • Availability
  • Add to Cart