COM448679871 Combi-Blocks, Inc.

4-(Di-m-tolyl-amino)-benzaldehyde

  • MDL number MFCD09025331
  • Cas Number : 332411-18-4
  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2458
Availability details for
Quantity
  • Availability
  • Add to Cart