COM448679883 Combi-Blocks, Inc.

4-(o-Tolyloxy)piperidine

  • MDL number MFCD06248344
  • Cas Number : 63843-42-5
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2480
Availability details for
Quantity
  • Availability
  • Add to Cart