COM448679891 Combi-Blocks, Inc.

4-(Pyridin-2-yloxy)benzaldehyde, HCl

  • MDL number MFCD03840160
  • Cas Number : 1172759-99-7
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2502
Availability details for
Quantity
  • Availability
  • Add to Cart