COM448679899 Combi-Blocks, Inc.

4-(Pyrrolidin-1-ylmethyl)-2(1h)-quinolinone

  • MDL number MFCD11226117
  • Cas Number : 1086393-55-6
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2516

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2516
Availability details for
Quantity
  • Availability
  • Add to Cart