COM448679923 Combi-Blocks, Inc.

tert-Butyl 2-bromobutyrate

  • MDL number MFCD00209634
  • Cas Number : 24457-21-4
  • Molecular Weight : 223.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2578
Availability details for
Quantity
  • Availability
  • Add to Cart