COM448679954 Combi-Blocks, Inc.

4-[(4-Methyl-benzylamino)-methyl]-3-nitro-benzoic acid methyl ester

  • MDL number MFCD03426385
  • Cas Number : 675602-57-0
  • Molecular Weight : 314.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2641

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2641
Availability details for
Quantity
  • Availability
  • Add to Cart