COM448679964 Combi-Blocks, Inc.

4-[3,5-Bis(trifluoromethyl)phenoxy]benzenesulfonyl chloride

  • MDL number MFCD01631871
  • Cas Number : 885950-90-3
  • Molecular Weight : 404.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2658
Availability details for
Quantity
  • Availability
  • Add to Cart