COM448679989 Combi-Blocks, Inc.

4-Amino-2-methoxy-5-pyrimidinecarbonitrile

  • MDL number MFCD00067828
  • Cas Number : 6964-55-2
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2737

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2737
Availability details for
Quantity
  • Availability
  • Add to Cart