COM448680003 Combi-Blocks, Inc.

Benzyl n-boc-4-isopropyl-4-piperidinecarboxylate

  • MDL number MFCD16495869
  • Cas Number : 1226776-80-2
  • Molecular Weight : 361.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2772

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2772
Availability details for
Quantity
  • Availability
  • Add to Cart