COM448680006 Combi-Blocks, Inc.

3-Amino-n-(4,5-dihydro-3h-pyrrol-2-yl)-benzenesulfonamide

  • MDL number MFCD03147395
  • Cas Number : 379255-71-7
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2775
Availability details for
Quantity
  • Availability
  • Add to Cart