COM448680011 Combi-Blocks, Inc.

2,3,5,6-Tetramethoxyisonicotinaldehyde

  • MDL number MFCD22887385
  • Cas Number : 1414864-14-4
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2790
Availability details for
Quantity
  • Availability
  • Add to Cart