COM448680021 Combi-Blocks, Inc.

4'-Aminophenacyl bromide

  • MDL number MFCD09744997
  • Cas Number : 23442-14-0
  • Molecular Weight : 214.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2813
Availability details for
Quantity
  • Availability
  • Add to Cart