COM448680032 Combi-Blocks, Inc.

4-Anilino-1-benzylpiperidine

  • MDL number MFCD00023738
  • Cas Number : 1155-56-2
  • Molecular Weight : 266.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2838
Availability details for
Quantity
  • Availability
  • Add to Cart