COM448680035 Combi-Blocks, Inc.

4-Benzenesulfonylamino-4-oxo-butyric acid

  • MDL number MFCD01861218
  • Cas Number : 100462-43-9
  • Molecular Weight : 257.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2845
Availability details for
Quantity
  • Availability
  • Add to Cart