COM448680038 Combi-Blocks, Inc.

4-Benzo[1,3]dioxol-5-yl-4-hydroxycyclohexanone

  • MDL number MFCD09800394
  • Cas Number : 150019-57-1
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2850
Availability details for
Quantity
  • Availability
  • Add to Cart