COM448680064 Combi-Blocks, Inc.

4-Boc-7-chloro-2,3,4,5-tetrahydro-1h-benzo[e][1,4]diazepine

  • MDL number MFCD07369868
  • Cas Number : 886364-33-6
  • Molecular Weight : 282.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2896
Availability details for
Quantity
  • Availability
  • Add to Cart