COM448680074 Combi-Blocks, Inc.

4-Bromo-1-(4-fluorophenyl)-1h-pyrazolo[3,4-c]pyridine

  • MDL number MFCD17169896
  • Cas Number : 1220165-54-7
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2908
Availability details for
Quantity
  • Availability
  • Add to Cart