COM448680081 Combi-Blocks, Inc.

4-Bromo-2-(tert-butyl)pyridine

  • MDL number MFCD11223211
  • Cas Number : 1086381-30-7
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2921

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2921
Availability details for
Quantity
  • Availability
  • Add to Cart