COM448680087 Combi-Blocks, Inc.

4-Bromo-3,3-dimethylindolin-2-one

  • MDL number MFCD18711494
  • Cas Number : 870552-47-9
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2928

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2928
Availability details for
Quantity
  • Availability
  • Add to Cart