COM448680091 Combi-Blocks, Inc.

4-Bromo-3-nitrobenzyl bromide

  • MDL number MFCD08059131
  • Cas Number : 326595-66-8
  • Molecular Weight : 294.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2933
Availability details for
Quantity
  • Availability
  • Add to Cart