COM448680104 Combi-Blocks, Inc.

4-Carboxymethoxy-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD02178989
  • Cas Number : 162504-85-0
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2958
Availability details for
Quantity
  • Availability
  • Add to Cart