COM448680123 Combi-Blocks, Inc.

4-Chloro-2-(pyrrolidin-1-yl)pyrimidine

  • MDL number MFCD09607721
  • Cas Number : 33852-01-6
  • Molecular Weight : 183.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2993
Availability details for
Quantity
  • Availability
  • Add to Cart