COM448680124 Combi-Blocks, Inc.

1-Boc-7-fluoro-1h-indazole

  • MDL number MFCD18911746
  • Cas Number : 1305320-59-5
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2994

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2994
Availability details for
Quantity
  • Availability
  • Add to Cart