COM448680156 Combi-Blocks, Inc.

4-Chloro-7-fluoro-quinoline-3-carbonitrile

  • MDL number MFCD06796584
  • Cas Number : 622369-70-4
  • Molecular Weight : 206.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3067
Availability details for
Quantity
  • Availability
  • Add to Cart