COM448680178 Combi-Blocks, Inc.

4-Cyano-1-n-fmoc-piperidine

  • MDL number MFCD02683059
  • Cas Number : 391248-16-1
  • Molecular Weight : 332.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3116

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3116
Availability details for
Quantity
  • Availability
  • Add to Cart