COM448680265 Combi-Blocks, Inc.

4-Methoxyindoline-2,3-dione

  • MDL number MFCD10000746
  • Cas Number : 108937-87-7
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3271

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3271
Availability details for
Quantity
  • Availability
  • Add to Cart