COM448680268 Combi-Blocks, Inc.

4-Methoxyphenylphosphonic acid diethyl ester

  • MDL number MFCD00015670
  • Cas Number : 3762-33-2
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3282
Availability details for
Quantity
  • Availability
  • Add to Cart