COM448680281 Combi-Blocks, Inc.

4-Methyl-2-pyrrolidin-1-ylmethyl-piperazine-1-carboxylic acid benzyl ester

  • MDL number MFCD03426445
  • Cas Number : 675602-56-9
  • Molecular Weight : 317.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3312
Availability details for
Quantity
  • Availability
  • Add to Cart